missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EGF / Epidermal Growth Factor Protein
A DNA sequence encoding the Homo sapiens (Human) EGF / Epidermal Growth Factor, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_001171601.1 |
---|---|
Gene ID (Entrez) | 1950 |
Name | EGF / Epidermal Growth Factor Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16055161
|
enQuireBio™
QP8537-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16075161
|
enQuireBio™
QP8537-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16065161
|
enQuireBio™
QP8537-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16085161
|
enQuireBio™
QP8537-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16095161
|
enQuireBio™
QP8537-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16005171
|
enQuireBio™
QP8537-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001171601.1 | |
EGF / Epidermal Growth Factor Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
1950 | |
Research Use Only | |
EGF | |
Recombinant Protein | |
E. coli | |
PLSHDGYCLH DGVCMYIEAL DKYACNCVVG YIGERCQYRD LKWWELR | |
Greater than 90% as determined by SDS-PAGE. |