missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EGF / Epidermal Growth Factor Protein
A DNA sequence encoding the Homo sapiens (Human) EGF / Epidermal Growth Factor, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8537-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001171601.1 | |
EGF / Epidermal Growth Factor Protein | |
Research Use Only | |
EGF | |
Recombinant Protein | |
E. coli | |
PLSHDGYCLH DGVCMYIEAL DKYACNCVVG YIGERCQYRD LKWWELR | |
Greater than 90% as determined by SDS-PAGE. |
1950 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |