missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DUSP3 Protein
A cDNA sequence encoding the DUSP3 was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | DUSP3 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | Specific activity: >1,750 units/mg. Enzymatic activity was confirmed by measuring the amount of enzyme hydrolyzing 1 nmole of p-nitrophenyl phosphate (pNPP) per minute at 37C, pH7.5 using 10mM of substrate. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15974308
|
enQuireBio™
QP10579-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15964308
|
enQuireBio™
QP10579-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15954308
|
enQuireBio™
QP10579-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
DUSP3 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSGSFELSVQ DLNDLLSDGS GCYSLPSQPC NEVTPRIYVG NASVAQDIPK LQKLGITHVL NAAEGRSFMH VNTNANFYKD SGITYLGIKA NDTQEFNLSA YFERAADFID QALAQKNGRV LVHCREGYSR SPTLVIAYLM MRQKMDVKSA LSIVRQNREI GPNDGFLAQL CQLNDRLAKE GKLKP | |
Greater than 95% as determined by SDS-PAGE. |
Research Use Only | |
Specific activity: >1,750 units/mg. Enzymatic activity was confirmed by measuring the amount of enzyme hydrolyzing 1 nmole of p-nitrophenyl phosphate (pNPP) per minute at 37C, pH7.5 using 10mM of substrate. | |
Human | |
His | |
The DUSP3 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 2mM DTT and 10% glycerol. |