missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DUSP3 Protein
A cDNA sequence encoding the DUSP3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10579-1mg
Additional Details : Weight : 0.01000kg
Specifications
DUSP3 Protein | |
Research Use Only | |
Specific activity: >1,750 units/mg. Enzymatic activity was confirmed by measuring the amount of enzyme hydrolyzing 1 nmole of p-nitrophenyl phosphate (pNPP) per minute at 37C, pH7.5 using 10mM of substrate. | |
Human | |
His | |
The DUSP3 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 2mM DTT and 10% glycerol. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSGSFELSVQ DLNDLLSDGS GCYSLPSQPC NEVTPRIYVG NASVAQDIPK LQKLGITHVL NAAEGRSFMH VNTNANFYKD SGITYLGIKA NDTQEFNLSA YFERAADFID QALAQKNGRV LVHCREGYSR SPTLVIAYLM MRQKMDVKSA LSIVRQNREI GPNDGFLAQL CQLNDRLAKE GKLKP | |
Greater than 95% as determined by SDS-PAGE. |