missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Draxin Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£327.21
Specifications
Gene ID (Entrez) | 374946 |
---|---|
Molecular Weight (g/mol) | 36.9 kDa |
Name | Draxin |
Quantity | 25μg |
Source | Mouse myeloma cell line,NS0-derived human Draxin protein Gly26-Val349,with a C-terminal 6-His tag |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18186150
|
R&D Systems™
6148-DR-025/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
£327.21
25µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18351428
|
R&D Systems™
6148-DR-025 |
£327.21
25µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Human Draxin Protein is derived from NS0. The Recombinant Human Draxin Protein has been validated for the following applications: Bioactivity.Specifications
374946 | |
Draxin | |
Mouse myeloma cell line,NS0-derived human Draxin protein Gly26-Val349,with a C-terminal 6-His tag | |
Monomer | |
Recombinant |
36.9 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |