missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Draxin Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 6148-DR-025
Additional Details : Weight : 0.00970kg
Description
The Recombinant Human Draxin Protein is derived from NS0. The Recombinant Human Draxin Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | |
36.9 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
374946 | |
Draxin | |
Mouse myeloma cell line,NS0-derived human Draxin protein Gly26-Val349,with a C-terminal 6-His tag | |
Monomer | |
Recombinant |