missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNA nucleotidylexotransferase Protein
A DNA sequence encoding the Homo sapiens (Human) DNA nucleotidylexotransferase, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_001017520.1 |
---|---|
Gene ID (Entrez) | 1791 |
Name | DNA nucleotidylexotransferase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15948829
|
enQuireBio™
QP5941-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15968829
|
enQuireBio™
QP5941-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15958829
|
enQuireBio™
QP5941-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15978829
|
enQuireBio™
QP5941-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15988829
|
enQuireBio™
QP5941-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15998829
|
enQuireBio™
QP5941-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001017520.1 | |
DNA nucleotidylexotransferase Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
1791 | |
Research Use Only | |
DNTT | |
Recombinant Protein | |
E. coli | |
MDPPRASHLS PRKKRPRQTG ALMASSPQDI KFQDLVVFIL EKKMGTTRRA FLMELARRKG FRVENELSDS VTHIVAENNS GSDVLEWLQA QKVQVSSQPE LLDVSWLIEC IRAGKPVEMT GKHQLVVRRD YSDSTNPGPP KTPPIAVQKI SQYACQRRTT LNNCNQIFTD AFDILAENCE FRENEDSCVT FMRAASVLKS LPFTIISMKD TEGIPCLGSK VKGIIEEIIE DGESSEVKAV LNDERYQSFK LFTSVFGVGL KTSEKWFRMG FRTLSKVRSD KSLKFTRMQK AGFLYYEDLV SCVTRAEAEA VSVLVKEAVW AFLPDAFVTM TGGFRRGKKM GHDVDFLITS PGSTEDEEQL LQKVMNLWEK KGLLLYYDLV ESTFEKLRLP SRKVDALDHF QKCFLIFKLP RQRVDSDQSS WQEGKTWKAI RVDLVLCPYE RRAFALLGWT GSRQFERDLR RYATHERKMI LDNHALYDKT KRIFLKAESE EEIFAHLGLD YIEPWERNA | |
Greater than 90% as determined by SDS-PAGE. |