missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNA nucleotidylexotransferase Protein
A DNA sequence encoding the Homo sapiens (Human) DNA nucleotidylexotransferase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5941-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001017520.1 | |
DNA nucleotidylexotransferase Protein | |
Research Use Only | |
DNTT | |
Recombinant Protein | |
E. coli | |
MDPPRASHLS PRKKRPRQTG ALMASSPQDI KFQDLVVFIL EKKMGTTRRA FLMELARRKG FRVENELSDS VTHIVAENNS GSDVLEWLQA QKVQVSSQPE LLDVSWLIEC IRAGKPVEMT GKHQLVVRRD YSDSTNPGPP KTPPIAVQKI SQYACQRRTT LNNCNQIFTD AFDILAENCE FRENEDSCVT FMRAASVLKS LPFTIISMKD TEGIPCLGSK VKGIIEEIIE DGESSEVKAV LNDERYQSFK LFTSVFGVGL KTSEKWFRMG FRTLSKVRSD KSLKFTRMQK AGFLYYEDLV SCVTRAEAEA VSVLVKEAVW AFLPDAFVTM TGGFRRGKKM GHDVDFLITS PGSTEDEEQL LQKVMNLWEK KGLLLYYDLV ESTFEKLRLP SRKVDALDHF QKCFLIFKLP RQRVDSDQSS WQEGKTWKAI RVDLVLCPYE RRAFALLGWT GSRQFERDLR RYATHERKMI LDNHALYDKT KRIFLKAESE EEIFAHLGLD YIEPWERNA | |
Greater than 90% as determined by SDS-PAGE. |
1791 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |