missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Desmoglein-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Desmoglein-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP10015-iv-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001933.2 | |
Desmoglein-1 Protein | |
Research Use Only | |
DSG1 | |
Recombinant Protein | |
E. coli | |
EWIKFAAACR EGEDNSKRNP IAKIHSDCAA NQQVTYRISG VGIDQPPYGI FVINQKTGEI NITSIVDREV TPFFIIYCRA LNSMGQDLER PLELRVRVLD INDNPPVFSM ATFAGQIEEN SNANTLVMIL NATDADEPNN LNSKIAFKII RQEPSDSPMF IINRNTGEIR TMNNFLDREQ YGQYALAVRG SDRDGGADGM SAECECNIKI LDVNDNIPYM EQSSYTIEIQ ENTLNSNLLE IRVIDLDEEF SANWMAVIFF ISGNEGNWFE IEMNERTNVG ILKVVKPLDY EAMQSLQLSI GVRNKAEFHH SIMSQYKLKA SAISVTVLNV IEGPVFRPGS KTYVVTGNMG SNDKVGDFVA TDLDTGRPST TVRYVMGNNP ADLLAVDSRT GKLTLKNKVT KEQYNMLGGK YQGTILSIDD NLQRTCTGTI NINIQSFGND DRTNTEPNTK ITTNTGRQES TSSTNYDTST TSTDSSQVYS SEPGNGAKDL LSDNVHFGP | |
Greater than 80% as determined by SDS-PAGE. |
1828 | |
100 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |