missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Desmoglein-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Desmoglein-1, was expressed in the hosts and tags indicated.
£428.07 - £1240.00
Specifications
Accession Number | NP_001933.2 |
---|---|
Gene ID (Entrez) | 1828 |
Name | Desmoglein-1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15942758
|
enQuireBio™
QP10015-IV-100UG |
100 μg |
£1240.00
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15962758
|
enQuireBio™
QP10015-IV-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15952758
|
enQuireBio™
QP10015-IV-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001933.2 | |
Desmoglein-1 Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
1828 | |
Research Use Only | |
DSG1 | |
Recombinant Protein | |
E. coli | |
EWIKFAAACR EGEDNSKRNP IAKIHSDCAA NQQVTYRISG VGIDQPPYGI FVINQKTGEI NITSIVDREV TPFFIIYCRA LNSMGQDLER PLELRVRVLD INDNPPVFSM ATFAGQIEEN SNANTLVMIL NATDADEPNN LNSKIAFKII RQEPSDSPMF IINRNTGEIR TMNNFLDREQ YGQYALAVRG SDRDGGADGM SAECECNIKI LDVNDNIPYM EQSSYTIEIQ ENTLNSNLLE IRVIDLDEEF SANWMAVIFF ISGNEGNWFE IEMNERTNVG ILKVVKPLDY EAMQSLQLSI GVRNKAEFHH SIMSQYKLKA SAISVTVLNV IEGPVFRPGS KTYVVTGNMG SNDKVGDFVA TDLDTGRPST TVRYVMGNNP ADLLAVDSRT GKLTLKNKVT KEQYNMLGGK YQGTILSIDD NLQRTCTGTI NINIQSFGND DRTNTEPNTK ITTNTGRQES TSSTNYDTST TSTDSSQVYS SEPGNGAKDL LSDNVHFGP | |
Greater than 80% as determined by SDS-PAGE. |