missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Desmin Protein

A cDNA sequence encoding the Desmin was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11662-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15917958

  • £3965.00 / 1mg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Desmin Protein
Research Use Only
Recombinant Protein
E. coli
MSQAYSSSQR VSSYRRTFGG APGFPLGSPL SSPVFPRAGF GSKGSSSSVT SRVYQVSRTS GGAGGLGSLR ASRLGTTRTP SSYGAGELLD FSLADAVNQE FLTTRTNEKV ELQELNDRFA NYIEKVRFLE QQNAALAAEV NRLKGREPTR VAELYEEELR ELRRQVEVLT NQRARVDVER DNLLDDLQRL KAKLQEEIQL KEEAENNLAA FRADVDAATL ARIDLERRIE SLNEEIAFLK KVHEEEIREL QAQLQEQQVQ VEMDMSKPDL TAALRDIRAQ YETIAAKNIS EAEEWYKSKV SDLTQAANKN NDALRQAKQE MMEYRHQIQS YTCEIDALKG TNDSLMRQMR ELEDRFASEA SGYQDNIARL EEEIRHLKDE MARHLREYQD LLNVKMALDV EIATYRKLLE GEESRINLPI QTYSALNFRE TSPEQRGSEV HTKKTVMIKT IETRDGEVVS EATQQQHEVL
Greater than 95.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
Desmin was lyophilized from a 1 mg/ml solution containing 30mM Tris-HCl pH 8.0, 9.5M urea, 2mM DTT, 2mM EDTA and 10mM methylammonium chloride.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers