missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Desmin Protein
A cDNA sequence encoding the Desmin was constructed and used to recombinantly synthesize the protein.
£99.15 - £3965.00
Specifications
Name | Desmin Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15937958
|
enQuireBio™
QP11662-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15927958
|
enQuireBio™
QP11662-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15917958
|
enQuireBio™
QP11662-1MG |
1 mg |
£3965.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Desmin Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
Desmin was lyophilized from a 1 mg/ml solution containing 30mM Tris-HCl pH 8.0, 9.5M urea, 2mM DTT, 2mM EDTA and 10mM methylammonium chloride. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MSQAYSSSQR VSSYRRTFGG APGFPLGSPL SSPVFPRAGF GSKGSSSSVT SRVYQVSRTS GGAGGLGSLR ASRLGTTRTP SSYGAGELLD FSLADAVNQE FLTTRTNEKV ELQELNDRFA NYIEKVRFLE QQNAALAAEV NRLKGREPTR VAELYEEELR ELRRQVEVLT NQRARVDVER DNLLDDLQRL KAKLQEEIQL KEEAENNLAA FRADVDAATL ARIDLERRIE SLNEEIAFLK KVHEEEIREL QAQLQEQQVQ VEMDMSKPDL TAALRDIRAQ YETIAAKNIS EAEEWYKSKV SDLTQAANKN NDALRQAKQE MMEYRHQIQS YTCEIDALKG TNDSLMRQMR ELEDRFASEA SGYQDNIARL EEEIRHLKDE MARHLREYQD LLNVKMALDV EIATYRKLLE GEESRINLPI QTYSALNFRE TSPEQRGSEV HTKKTVMIKT IETRDGEVVS EATQQQHEVL | |
Greater than 95.0% as determined by SDS-PAGE. |