missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CSTA Protein
A cDNA sequence encoding the CSTA was constructed and used to recombinantly synthesize the protein.
£211.00 - £568.00
Specifications
Name | CSTA Protein |
---|---|
Product Type | Recombinant Protein |
Protein Tag | GST |
Host Species | E. coli |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15947598
|
enQuireBio™
QP11547-GST-2UG |
2 μg |
£211.00
2µg |
Estimated Shipment: 14-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
15957598
|
enQuireBio™
QP11547-GST-5UG |
5 μg |
£315.00
5µg |
Estimated Shipment: 14-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
15937598
|
enQuireBio™
QP11547-GST-10UG |
10 μg |
£568.00
10µg |
Estimated Shipment: 14-09-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
Specifications
CSTA Protein | |
GST | |
Human | |
1475 |
Recombinant Protein | |
E. coli | |
MIPGGLSEAK PATPEIQEIV DKVKPQLEEK TNETYGKLEA VQYKTQVVAG TNYYIKVRAG DNKYMHLKVF KSLPGQNEDL VLTGYQVDKN KDDELTGF | |
CSTA |