missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CSTA Protein

A cDNA sequence encoding the CSTA was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11547-GST-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15937598

  • £540.00 / 10µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

1475
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MIPGGLSEAK PATPEIQEIV DKVKPQLEEK TNETYGKLEA VQYKTQVVAG TNYYIKVRAG DNKYMHLKVF KSLPGQNEDL VLTGYQVDKN KDDELTGF
CSTA Protein
Research Use Only
CSTA
Human
GST
Cystatin-A in 50mM Tris-Acetate, pH 7.5, 1mM EDTA and 20% Glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers