missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CSNK1A1 / CKI-alpha / CK1 Protein
A DNA sequence encoding the Homo sapiens (Human) CSNK1A1 / CKI-alpha / CK1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5879-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001020276.1 | |
CSNK1A1 / CKI-alpha / CK1 Protein | |
Research Use Only | |
CSNK1A1 | |
Recombinant Protein | |
E. coli | |
ASSSGSKAEF IVGGKYKLVR KIGSGSFGDI YLAINITNGE EVAVKLESQK ARHPQLLYES KLYKILQGGV GIPHIRWYGQ EKDYNVLVMD LLGPSLEDLF NFCSRRFTMK TVLMLADQMI SRIEYVHTKN FIHRDIKPDN FLMGIGRHCN KLFLIDFGLA KKYRDNRTRQ HIPYREDKNL TGTARYASIN AHLGIEQSRR DDMESLGYVL MYFNRTSLPW QGLKAATKKQ KYEKISEKKM STPVEVLCKG FPAEFAMYLN YCRGLRFEEA PDYMYLRQLF RILFRTLNHQ YDYTFDWTML KQKAAQQAAS SSGQGQQAQT PTGKQTDKTK SNMKGF | |
Greater than 90% as determined by SDS-PAGE. |
1452 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |