missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CSNK1A1 / CKI-alpha / CK1 Protein
A DNA sequence encoding the Homo sapiens (Human) CSNK1A1 / CKI-alpha / CK1, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_001020276.1 |
---|---|
Gene ID (Entrez) | 1452 |
Name | CSNK1A1 / CKI-alpha / CK1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15958399
|
enQuireBio™
QP5879-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15978399
|
enQuireBio™
QP5879-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15968399
|
enQuireBio™
QP5879-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15988399
|
enQuireBio™
QP5879-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15998399
|
enQuireBio™
QP5879-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15908409
|
enQuireBio™
QP5879-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001020276.1 | |
CSNK1A1 / CKI-alpha / CK1 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
1452 | |
Research Use Only | |
CSNK1A1 | |
Recombinant Protein | |
E. coli | |
ASSSGSKAEF IVGGKYKLVR KIGSGSFGDI YLAINITNGE EVAVKLESQK ARHPQLLYES KLYKILQGGV GIPHIRWYGQ EKDYNVLVMD LLGPSLEDLF NFCSRRFTMK TVLMLADQMI SRIEYVHTKN FIHRDIKPDN FLMGIGRHCN KLFLIDFGLA KKYRDNRTRQ HIPYREDKNL TGTARYASIN AHLGIEQSRR DDMESLGYVL MYFNRTSLPW QGLKAATKKQ KYEKISEKKM STPVEVLCKG FPAEFAMYLN YCRGLRFEEA PDYMYLRQLF RILFRTLNHQ YDYTFDWTML KQKAAQQAAS SSGQGQQAQT PTGKQTDKTK SNMKGF | |
Greater than 90% as determined by SDS-PAGE. |