missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CSF2RA / GM-CSFR / CD116 Protein
A DNA sequence encoding the Homo sapiens (Human) CSF2RA / GM-CSFR / CD116, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_001155001.1 |
---|---|
Gene ID (Entrez) | 1438 |
Name | CSF2RA / GM-CSFR / CD116 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15938389
|
enQuireBio™
QP5877-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15958389
|
enQuireBio™
QP5877-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15948389
|
enQuireBio™
QP5877-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15968389
|
enQuireBio™
QP5877-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15978389
|
enQuireBio™
QP5877-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15988389
|
enQuireBio™
QP5877-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001155001.1 | |
CSF2RA / GM-CSFR / CD116 Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
1438 | |
Research Use Only | |
CSF2RA | |
Recombinant Protein | |
E. coli | |
EKSDLRTVAP ASSLNVRFDS RTMNLSWDCQ ENTTFSKCFL TDKKNRVVEP RLSNNECSCT FREICLHEGV TFEVHVNTSQ RGFQQKLLYP NSGREGTAAQ NFSCFIYNAD LMNCTWARGP TAPRDVQYFL YIRNSKRRRE IRCPYYIQDS GTHVGCHLDN LSGLTSRNYF LVNGTSREIG IQFFDSLLDT KKIERFNPPS NVTVRCNTTH CLVRWKQPRT YQKLSYLDFQ YQLDVHRKNT QPGTENLLIN VSGDLENRYN FPSSEPRAKH SVKIRAADVR ILNWSSWSEA IEFGSDDG | |
Greater than 90% as determined by SDS-PAGE. |