missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CSF2RA / GM-CSFR / CD116 Protein
A DNA sequence encoding the Homo sapiens (Human) CSF2RA / GM-CSFR / CD116, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5877-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001155001.1 | |
CSF2RA / GM-CSFR / CD116 Protein | |
Research Use Only | |
CSF2RA | |
Recombinant Protein | |
E. coli | |
EKSDLRTVAP ASSLNVRFDS RTMNLSWDCQ ENTTFSKCFL TDKKNRVVEP RLSNNECSCT FREICLHEGV TFEVHVNTSQ RGFQQKLLYP NSGREGTAAQ NFSCFIYNAD LMNCTWARGP TAPRDVQYFL YIRNSKRRRE IRCPYYIQDS GTHVGCHLDN LSGLTSRNYF LVNGTSREIG IQFFDSLLDT KKIERFNPPS NVTVRCNTTH CLVRWKQPRT YQKLSYLDFQ YQLDVHRKNT QPGTENLLIN VSGDLENRYN FPSSEPRAKH SVKIRAADVR ILNWSSWSEA IEFGSDDG | |
Greater than 90% as determined by SDS-PAGE. |
1438 | |
200 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |