missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Contactin-associated protein 1
A DNA sequence encoding the Homo sapiens (Human) Contactin-associated protein 1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5850-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003623.1 | |
Contactin-associated protein 1 | |
Research Use Only | |
CNTNAP1 | |
Recombinant Protein | |
E. coli | |
DEELVGPLYA RSLGASSYYS LLTAPRFARL HGISGWSPRI GDPNPWLQID LMKKHRIRAV ATQGSFNSWD WVTRYMLLYG DRVDSWTPFY QRGHNSTFFG NVNESAVVRH DLHFHFTARY IRIVPLAWNP RGKIGLRLGL YGCPYKADIL YFDGDDAISY RFPRGVSRSL WDVFAFSFKT EEKDGLLLHA EGAQGDYVTL ELEGAHLLLH MSLGSSPIQP RPGHTTVSAG GVLNDQHWHY VRVDRFGRDV NFTLDGYVQR FILNGDFERL NLDTEMFIGG LVGAARKNLA YRHNFRGCIE NVIFNRVNIA DLAVRRHSRI TFEGKVAFRC L | |
Greater than 90% as determined by SDS-PAGE. |
8506 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |