missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Contactin-associated protein 1
A DNA sequence encoding the Homo sapiens (Human) Contactin-associated protein 1, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_003623.1 |
---|---|
Gene ID (Entrez) | 8506 |
Name | Contactin-associated protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15928219
|
enQuireBio™
QP5850-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15948219
|
enQuireBio™
QP5850-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15938219
|
enQuireBio™
QP5850-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15958219
|
enQuireBio™
QP5850-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15968219
|
enQuireBio™
QP5850-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15978219
|
enQuireBio™
QP5850-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_003623.1 | |
Contactin-associated protein 1 | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
8506 | |
Research Use Only | |
CNTNAP1 | |
Recombinant Protein | |
E. coli | |
DEELVGPLYA RSLGASSYYS LLTAPRFARL HGISGWSPRI GDPNPWLQID LMKKHRIRAV ATQGSFNSWD WVTRYMLLYG DRVDSWTPFY QRGHNSTFFG NVNESAVVRH DLHFHFTARY IRIVPLAWNP RGKIGLRLGL YGCPYKADIL YFDGDDAISY RFPRGVSRSL WDVFAFSFKT EEKDGLLLHA EGAQGDYVTL ELEGAHLLLH MSLGSSPIQP RPGHTTVSAG GVLNDQHWHY VRVDRFGRDV NFTLDGYVQR FILNGDFERL NLDTEMFIGG LVGAARKNLA YRHNFRGCIE NVIFNRVNIA DLAVRRHSRI TFEGKVAFRC L | |
Greater than 90% as determined by SDS-PAGE. |