missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CKMBITI Protein
A cDNA sequence encoding the CKMBITI was constructed and used to recombinantly synthesize the protein.
£99.15 - £1320.00
Specifications
Name | CKMBITI Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | 486 IU/mg at 37C. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15934228
|
enQuireBio™
QP10553-10UG |
10 μg |
£99.15
10µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15954228
|
enQuireBio™
QP10553-50UG |
50 μg |
£155.00
50µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15944228
|
enQuireBio™
QP10553-1MG |
1 mg |
£1320.00
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CKMBITI Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
P. pastoris | |
The 7.9 mg/ml protein solution contains 0.02M Potassium Phosphate, 1mM DTT and 50% glycerol, pH 5.0-6.0. |
Research Use Only | |
486 IU/mg at 37C. | |
Human | |
Untagged | |
Greater than 95.0% as determined by SDS-PAGE. |