missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CKMBITI Protein
A cDNA sequence encoding the CKMBITI was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10553-50ug
Additional Details : Weight : 0.01000kg
Specifications
CKMBITI Protein | |
Research Use Only | |
486 IU/mg at 37C. | |
Human | |
Untagged | |
Greater than 95.0% as determined by SDS-PAGE. |
50 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
P. pastoris | |
The 7.9 mg/ml protein solution contains 0.02M Potassium Phosphate, 1mM DTT and 50% glycerol, pH 5.0-6.0. |