missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CHI3L1 / YKL40 Protein
A DNA sequence encoding the Homo sapiens (Human) CHI3L1 / YKL40, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5829-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001267.2 | |
CHI3L1 / YKL40 Protein | |
Research Use Only | |
CHI3L1 | |
Recombinant Protein | |
E. coli | |
YKLVCYYTSW SQYREGDGSC FPDALDRFLC THIIYSFANI SNDHIDTWEW NDVTLYGMLN TLKNRNPNLK TLLSVGGWNF GSQRFSKIAS NTQSRRTFIK SVPPFLRTHG FDGLDLAWLY PGRRDKQHFT TLIKEMKAEF IKEAQPGKKQ LLLSAALSAG KVTIDSSYDI AKISQHLDFI SIMTYDFHGA WRGTTGHHSP LFRGQEDASP DRFSNTDYAV GYMLRLGAPA SKLVMGIPTF GRSFTLASSE TGVGAPISGP GIPGRFTKEA GTLAYYEICD FLRGATVHRI LGQQVPYATK GNQWVGYDDQ ESVKSKVQYL KDRQLAGAMV WALDLDDFQG SFCGQDLRFP LTNAIKDALA AT | |
Greater than 80% as determined by SDS-PAGE. |
1116 | |
1 mg | |
Not determined. | |
Full Length of Mature Protein | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |