missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CHI3L1 / YKL40 Protein

A DNA sequence encoding the Homo sapiens (Human) CHI3L1 / YKL40, was expressed in the hosts and tags indicated.

£145.46 - £1735.00

Specifications

Accession Number NP_001267.2
Gene ID (Entrez) 1116
Name CHI3L1 / YKL40 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15958069
View Documents Promotion Details
enQuireBio™
QP5829-EC-100UG
100 μg
£384.00
100µg
Ver productos alternativos
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 24-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15978069
missing translation for 'viewDocuments' Detalles de la promoción
enQuireBio™
QP5829-EC-1MG
1 mg
£1735.00
1mg
Ver productos alternativos
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 24-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15968069
missing translation for 'viewDocuments' Detalles de la promoción
enQuireBio™
QP5829-EC-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A Ver productos alternativos
missing translation for 'toSaveUpto' 
»
15988069
missing translation for 'viewDocuments' Detalles de la promoción
enQuireBio™
QP5829-EC-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A Ver productos alternativos
missing translation for 'toSaveUpto' 
»
15998069
missing translation for 'viewDocuments' Detalles de la promoción
enQuireBio™
QP5829-EC-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A Ver productos alternativos
missing translation for 'toSaveUpto' 
»
15908079
missing translation for 'viewDocuments' Detalles de la promoción
enQuireBio™
QP5829-EC-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A Ver productos alternativos
missing translation for 'toSaveUpto' 
»
Descripción

Descripción

Especificaciones

Especificaciones

NP_001267.2
CHI3L1 / YKL40 Protein
Not determined.
Full Length of Mature Protein
Human
His-SUMO
Tris-based buffer, 50% glycerol
1116
Research Use Only
CHI3L1
Recombinant Protein
E. coli
YKLVCYYTSW SQYREGDGSC FPDALDRFLC THIIYSFANI SNDHIDTWEW NDVTLYGMLN TLKNRNPNLK TLLSVGGWNF GSQRFSKIAS NTQSRRTFIK SVPPFLRTHG FDGLDLAWLY PGRRDKQHFT TLIKEMKAEF IKEAQPGKKQ LLLSAALSAG KVTIDSSYDI AKISQHLDFI SIMTYDFHGA WRGTTGHHSP LFRGQEDASP DRFSNTDYAV GYMLRLGAPA SKLVMGIPTF GRSFTLASSE TGVGAPISGP GIPGRFTKEA GTLAYYEICD FLRGATVHRI LGQQVPYATK GNQWVGYDDQ ESVKSKVQYL KDRQLAGAMV WALDLDDFQG SFCGQDLRFP LTNAIKDALA AT
Greater than 80% as determined by SDS-PAGE.
Videos
SDS
Documentos

Documentos

Certificados