missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cell growth-regulating nucleolar Protein

A DNA sequence encoding the Homo sapiens (Human) Cell growth-regulating nucleolar protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9912-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16088981

  • £1600.00 / 500µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001139197.1
Cell growth-regulating nucleolar Protein
Research Use Only
LYAR
Recombinant Protein
Yeast
MVFFTCNACG ESVKKIQVEK HVSVCRNCEC LSCIDCGKDF WGDDYKNHVK CISEDQKYGG KGYEGKTHKG DIKQQAWIQK ISELIKRPNV SPKVRELLEQ ISAFDNVPRK KAKFQNWMKN SLKVHNESIL DQVWNIFSEA SNSEPVNKEQ DQRPLHPVAN PHAEISTKVP ASKVKDAVEQ QGEVKKNKRE RKEERQKKRK REKKELKLEN HQENSRNQKP KKRKKGQEAD LEAGGEEVPE ANGSAGKRSK KKKQRKDSAS EEEAHVGAGK RKRRHSEVET DSKKKKMKLP EHPEGGEPED DEAPAKGKFN WKGTIKAILK QAPDNEITIK KLRKKVLAQY YTVTDEHHRS EEELLVIFNK KISKNPTFKL LKDKVKLVK
Greater than 90% as determined by SDS-PAGE.
55646
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers