missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cell growth-regulating nucleolar Protein
A DNA sequence encoding the Homo sapiens (Human) Cell growth-regulating nucleolar protein, was expressed in the hosts and tags indicated.
£223.39 - £2410.48
Specifications
Accession Number | NP_001139197.1 |
---|---|
Gene ID (Entrez) | 55646 |
Name | Cell growth-regulating nucleolar Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16048981
|
enQuireBio™
QP9912-YE-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16088981
|
enQuireBio™
QP9912-YE-500UG |
500 μg |
£1600.00
500µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16058981
|
enQuireBio™
QP9912-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16078981
|
enQuireBio™
QP9912-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16098981
|
enQuireBio™
QP9912-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16068981
|
enQuireBio™
QP9912-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001139197.1 | |
Cell growth-regulating nucleolar Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
55646 | |
Research Use Only | |
LYAR | |
Recombinant Protein | |
Yeast | |
MVFFTCNACG ESVKKIQVEK HVSVCRNCEC LSCIDCGKDF WGDDYKNHVK CISEDQKYGG KGYEGKTHKG DIKQQAWIQK ISELIKRPNV SPKVRELLEQ ISAFDNVPRK KAKFQNWMKN SLKVHNESIL DQVWNIFSEA SNSEPVNKEQ DQRPLHPVAN PHAEISTKVP ASKVKDAVEQ QGEVKKNKRE RKEERQKKRK REKKELKLEN HQENSRNQKP KKRKKGQEAD LEAGGEEVPE ANGSAGKRSK KKKQRKDSAS EEEAHVGAGK RKRRHSEVET DSKKKKMKLP EHPEGGEPED DEAPAKGKFN WKGTIKAILK QAPDNEITIK KLRKKVLAQY YTVTDEHHRS EEELLVIFNK KISKNPTFKL LKDKVKLVK | |
Greater than 90% as determined by SDS-PAGE. |