missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CEL Protein
A cDNA sequence encoding the CEL was constructed and used to recombinantly synthesize the protein.
£99.15 - £1320.00
Specifications
Gene ID (Entrez) | 1056 |
---|---|
Name | CEL Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | CEL |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15987098
|
enQuireBio™
QP11385-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15967098
|
enQuireBio™
QP11385-100UG |
100 μg |
£1320.00
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15977098
|
enQuireBio™
QP11385-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
1056 | |
Research Use Only | |
CEL | |
Human | |
Untagged | |
CEL protein filtered (0.4microm) and lyophilized in 0.5 mg/ml in 0.05M phosphate buffer and 0.075M NaCl pH 7.4. |
CEL Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Human | |
AKLGAVYTEG GFVEGVNKKL GLLGDSVDIF KGIPFAAPTK ALENPQPHPG WQGTLKAKNF KKRCLQATIT QDSTYGDEDC LYLNIWVPQG RKQVSRDLPV MIWIYGGAFL MGSGHGANFL NNYLYDGEEI ATRGNVIVVT FNYRVGPLGF LSTGDANLPG NYGLRDQHMA IAWVKRNIAA FGGDPNNITL FGESAGGASV SLQTLSPYNK GLIRRAISQS GVALSPWVIQ KNPLFWAKKV AEKVGCPVGD AARMAQCLKV TDPRALTLAY KVPLAGLEYP MLHYVGFVPV IDGDFIPADP INLYANAADI DYIAGTNNMD GHIFASIDMP AINKGNKKVT EEDFYKLVSE FTITKGLRGA KTTFDVYTES WAQDPSQENK KKTVVDFETD VLFLVPTEIA LAQHRANAKS AKTYAYLFSH PSRMPVYPKW VGADHADDIQ YVFGKPFATP TGYRPQDRTV SKAMIAYWTN FAKTGDPNMG DSAVPTHWEP YTTENSGYLE ITKKMGSSSM KRSLRTNFLR YWTLTYLALP TVTDQEATPV PPTGDSEATP VPPTGDSETA PVPPTGDSGA PPVPPTGDSG APPVPPTGDS GAPPVPPTGD SGAPPVPPTG DSGAPPVPPT GDSGAPPVPP TGDSGAPPVP PTGDSGAPPV PPTGDAGPPP VPPTGDSGAP PVPPTGDSGA PPVTPTGDSE TAPVPPTGDS GAPPVPPTGD SEAAPVPPTD DSKEAQMPAV IRF | |
Greater than 95.0% as determined by SDS-PAGE. |