missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CEL Protein
A cDNA sequence encoding the CEL was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11385-2ug
Additional Details : Weight : 0.01000kg
Specifications
1056 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Human | |
AKLGAVYTEG GFVEGVNKKL GLLGDSVDIF KGIPFAAPTK ALENPQPHPG WQGTLKAKNF KKRCLQATIT QDSTYGDEDC LYLNIWVPQG RKQVSRDLPV MIWIYGGAFL MGSGHGANFL NNYLYDGEEI ATRGNVIVVT FNYRVGPLGF LSTGDANLPG NYGLRDQHMA IAWVKRNIAA FGGDPNNITL FGESAGGASV SLQTLSPYNK GLIRRAISQS GVALSPWVIQ KNPLFWAKKV AEKVGCPVGD AARMAQCLKV TDPRALTLAY KVPLAGLEYP MLHYVGFVPV IDGDFIPADP INLYANAADI DYIAGTNNMD GHIFASIDMP AINKGNKKVT EEDFYKLVSE FTITKGLRGA KTTFDVYTES WAQDPSQENK KKTVVDFETD VLFLVPTEIA LAQHRANAKS AKTYAYLFSH PSRMPVYPKW VGADHADDIQ YVFGKPFATP TGYRPQDRTV SKAMIAYWTN FAKTGDPNMG DSAVPTHWEP YTTENSGYLE ITKKMGSSSM KRSLRTNFLR YWTLTYLALP TVTDQEATPV PPTGDSEATP VPPTGDSETA PVPPTGDSGA PPVPPTGDSG APPVPPTGDS GAPPVPPTGD SGAPPVPPTG DSGAPPVPPT GDSGAPPVPP TGDSGAPPVP PTGDSGAPPV PPTGDAGPPP VPPTGDSGAP PVPPTGDSGA PPVTPTGDSE TAPVPPTGDS GAPPVPPTGD SEAAPVPPTD DSKEAQMPAV IRF | |
Greater than 95.0% as determined by SDS-PAGE. |
CEL Protein | |
Research Use Only | |
CEL | |
Human | |
Untagged | |
CEL protein filtered (0.4microm) and lyophilized in 0.5 mg/ml in 0.05M phosphate buffer and 0.075M NaCl pH 7.4. |