missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cdc42 / Rac Protein
A DNA sequence encoding the Homo sapiens (Human) Cdc42/Rac, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8204-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_065074.1 | |
Cdc42 / Rac Protein | |
Research Use Only | |
PAK7 | |
Recombinant Protein | |
E. coli | |
SRVSHEQFRA ALQLVVSPGD PREYLANFIK IGEGSTGIVC IATEKHTGKQ VAVKKMDLRK QQRRELLFNE VVIMRDYHHD NVVDMYSSYL VGDELWVVME FLEGGALTDI VTHTRMNEEQ IATVCLSVLR ALSYLHNQGV IHRDIKSDSI LLTSDGRIKL SDFGFCAQVS KEVPKRKSLV GTPYWMAPEV ISRLPYGTEV DIWSLGIMVI EMIDGEPPYF NEPPLQAMRR IRDSLPPRVK DLHKVSSVLR GFLDLMLVRE PSQRATAQEL LGHPFLKLAG PPSCIVPLMR QYRHH | |
Greater than 90% as determined by SDS-PAGE. |
57144 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |