missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cdc42 / Rac Protein
A DNA sequence encoding the Homo sapiens (Human) Cdc42/Rac, was expressed in the hosts and tags indicated.
£145.46 - £1688.38
Specifications
Accession Number | NP_065074.1 |
---|---|
Gene ID (Entrez) | 57144 |
Name | Cdc42 / Rac Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16013261
|
enQuireBio™
QP8204-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16053261
|
enQuireBio™
QP8204-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16003261
|
enQuireBio™
QP8204-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16033261
|
enQuireBio™
QP8204-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16043261
|
enQuireBio™
QP8204-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16023261
|
enQuireBio™
QP8204-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_065074.1 | |
Cdc42 / Rac Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
57144 | |
Research Use Only | |
PAK7 | |
Recombinant Protein | |
E. coli | |
SRVSHEQFRA ALQLVVSPGD PREYLANFIK IGEGSTGIVC IATEKHTGKQ VAVKKMDLRK QQRRELLFNE VVIMRDYHHD NVVDMYSSYL VGDELWVVME FLEGGALTDI VTHTRMNEEQ IATVCLSVLR ALSYLHNQGV IHRDIKSDSI LLTSDGRIKL SDFGFCAQVS KEVPKRKSLV GTPYWMAPEV ISRLPYGTEV DIWSLGIMVI EMIDGEPPYF NEPPLQAMRR IRDSLPPRVK DLHKVSSVLR GFLDLMLVRE PSQRATAQEL LGHPFLKLAG PPSCIVPLMR QYRHH | |
Greater than 90% as determined by SDS-PAGE. |