missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD33 Protein
A cDNA sequence encoding the CD33 was constructed and used to recombinantly synthesize the protein.
£104.00 - £6030.00
Specifications
Name | CD33 Protein |
---|---|
Product Type | Recombinant Protein |
Protein Tag | His |
Host Species | Insect |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15976868
|
enQuireBio™
QP11327-HIS-2UG |
2 μg |
£104.00
2µg |
Estimated Shipment: 04-08-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
15986868
|
enQuireBio™
QP11327-HIS-5UG |
5 μg |
£104.00
5µg |
Estimated Shipment: 04-08-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
15956868
|
enQuireBio™
QP11327-HIS-10UG |
10 μg |
£159.00
10µg |
Estimated Shipment: 04-08-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
15966868
|
enQuireBio™
QP11327-HIS-20UG |
20 μg |
£159.00
20µg |
Estimated Shipment: 04-08-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
15906878
|
enQuireBio™
QP11327-HIS-INSECT-1MG |
1 mg |
£6030.00
1mg |
Estimated Shipment: 04-08-2022 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! |
Specifications
CD33 Protein | |
His | |
Human | |
Greater than 90% as determined by SDS-PAGE. | |
CD33 |
Recombinant Protein | |
Insect | |
MGSSHHHHHH SSGLVPRGSH MGSDPNFWLQ VQESVTVQEG LCVLVPCTFF HPIPYYDKNS PVHGYWFREG AIISGDSPVA TNKLDQEVQE ETQGRFRLLG DPSRNNCSLS IVDARRRDNG SYFFRMERGS TKYSYKSPQL SVHVTDLTHR PKILIPGTLE PGHSKNLTCS VSWACEQGTP PIFSWLSAAP TSLGPRTTHS SVLIITPRPQ DHGTNLTCQV KFAGAGVTTE RTIQLNVTYV PQNPTTGIFP GDGSGKQETR AGVVH | |
945 |