missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD33 Protein
A cDNA sequence encoding the CD33 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11327-HIS-20ug
Additional Details : Weight : 0.01000kg
Specifications
945 | |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
MGSSHHHHHH SSGLVPRGSH MGSDPNFWLQ VQESVTVQEG LCVLVPCTFF HPIPYYDKNS PVHGYWFREG AIISGDSPVA TNKLDQEVQE ETQGRFRLLG DPSRNNCSLS IVDARRRDNG SYFFRMERGS TKYSYKSPQL SVHVTDLTHR PKILIPGTLE PGHSKNLTCS VSWACEQGTP PIFSWLSAAP TSLGPRTTHS SVLIITPRPQ DHGTNLTCQV KFAGAGVTTE RTIQLNVTYV PQNPTTGIFP GDGSGKQETR AGVVH | |
Greater than 90% as determined by SDS-PAGE. |
CD33 Protein | |
Research Use Only | |
CD33 | |
Human | |
His | |
CD33 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea, and 10% glycerol. |