missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CD300E Protein

A cDNA sequence encoding the CD300E was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11326-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15936868

  • £155.00 / 20µg
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

342510
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSLKGPGSV TGTAGDSLTV WCQYESMYKG YNKYWCRGQY DTSCESIVET KGEEKVERNG RVSIRDHPEA LAFTVTMQNL NEDDAGSYWC KIQTVWVLDS WSRDPSDLVR VYVSPAITTP RRTTHPATPP IFLVVNPGRN LSTGEVLTQN SGFRLSSPH
Greater than 90.0% as determined by SDS-PAGE.
CD300E Protein
Research Use Only
CD300E
Human
His
CD300E protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers