missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD300E Protein
A cDNA sequence encoding the CD300E was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Gene ID (Entrez) | 342510 |
---|---|
Name | CD300E Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | CD300E |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15946868
|
enQuireBio™
QP11326-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15936868
|
enQuireBio™
QP11326-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15926868
|
enQuireBio™
QP11326-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
342510 | |
Research Use Only | |
CD300E | |
Human | |
His | |
CD300E protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0). |
CD300E Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSLKGPGSV TGTAGDSLTV WCQYESMYKG YNKYWCRGQY DTSCESIVET KGEEKVERNG RVSIRDHPEA LAFTVTMQNL NEDDAGSYWC KIQTVWVLDS WSRDPSDLVR VYVSPAITTP RRTTHPATPP IFLVVNPGRN LSTGEVLTQN SGFRLSSPH | |
Greater than 90.0% as determined by SDS-PAGE. |