missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD300E Protein
A cDNA sequence encoding the CD300E was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11326-1mg
Additional Details : Weight : 0.01000kg
Specifications
342510 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSLKGPGSV TGTAGDSLTV WCQYESMYKG YNKYWCRGQY DTSCESIVET KGEEKVERNG RVSIRDHPEA LAFTVTMQNL NEDDAGSYWC KIQTVWVLDS WSRDPSDLVR VYVSPAITTP RRTTHPATPP IFLVVNPGRN LSTGEVLTQN SGFRLSSPH | |
Greater than 90.0% as determined by SDS-PAGE. |
CD300E Protein | |
Research Use Only | |
CD300E | |
Human | |
His | |
CD300E protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0). |