missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD300C Protein
A cDNA sequence encoding the CD300C was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11325-HIS-INSECT-5ug
Additional Details : Weight : 0.01000kg
Specifications
10871 | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
MGSSHHHHHH SSGLVPRGSH MGSGYFPLSH PMTVAGPVGG SLSVQCRYEK EHRTLNKFWC RPPQILRCDK IVETKGSAGK RNGRVSIRDS PANLSFTVTL ENLTEEDAGT YWCGVDTPWL RDFHDPIVEV EVSVFPAGTT TASSPQSSMG TSGPPTKLPV HTWPSVTRKD SPEPSPHPGSLFSNVR | |
Greater than 85.0% as determined by SDS-PAGE. |
CD300C Protein | |
Research Use Only | |
CD300C | |
Human | |
His | |
CD300C protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol. |