missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD300C Protein
A cDNA sequence encoding the CD300C was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Gene ID (Entrez) | 10871 |
---|---|
Name | CD300C Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | CD300C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15976858
|
enQuireBio™
QP11325-HIS-1UG |
1 μg |
£99.15
1µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15916868
|
enQuireBio™
QP11325-HIS-INSECT-5UG |
5 μg |
£155.00
5µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15986858
|
enQuireBio™
QP11325-HIS-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15996858
|
enQuireBio™
QP11325-HIS-50UG |
50 μg |
£1320.00
50µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15966858
|
enQuireBio™
QP11325-HIS-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
10871 | |
Research Use Only | |
CD300C | |
Human | |
His | |
CD300C protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol. |
CD300C Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
MGSSHHHHHH SSGLVPRGSH MGSGYFPLSH PMTVAGPVGG SLSVQCRYEK EHRTLNKFWC RPPQILRCDK IVETKGSAGK RNGRVSIRDS PANLSFTVTL ENLTEEDAGT YWCGVDTPWL RDFHDPIVEV EVSVFPAGTT TASSPQSSMG TSGPPTKLPV HTWPSVTRKD SPEPSPHPGSLFSNVR | |
Greater than 85.0% as determined by SDS-PAGE. |