missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD200 Protein
A cDNA sequence encoding the CD200 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11312-HIS-10ug
Additional Details : Weight : 0.01000kg
Specifications
4345 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSQVQVVTQ DEREQLYTPA SLKCSLQNAQ EALIVTWQKK KAVSPENMVT FSENHGVVIQ PAYKDKINIT QLGLQNSTIT FWNITLEDEG CYMCLFNTFG FGKISGTACL TVYVQPIVSL HYKFSEDHLN ITCSATARPA PMVFWKVPRS GIENSTVTLS HPNGTTSVTS ILHIKDPKNQ VGKEVICQVL HLGTVTDFKQ TVNKG | |
Greater than 90% as determined by SDS-PAGE. |
CD200 Protein | |
Research Use Only | |
CD200 | |
Human | |
His | |
CD200 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |