missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD200 Protein
A cDNA sequence encoding the CD200 was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Gene ID (Entrez) | 4345 |
---|---|
Name | CD200 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | CD200 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15916808
|
enQuireBio™
QP11312-HIS-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15926808
|
enQuireBio™
QP11312-HIS-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15996798
|
enQuireBio™
QP11312-HIS-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15906808
|
enQuireBio™
QP11312-HIS-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15936808
|
enQuireBio™
QP11312-HIS-EC-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
4345 | |
Research Use Only | |
CD200 | |
Human | |
His | |
CD200 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |
CD200 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSQVQVVTQ DEREQLYTPA SLKCSLQNAQ EALIVTWQKK KAVSPENMVT FSENHGVVIQ PAYKDKINIT QLGLQNSTIT FWNITLEDEG CYMCLFNTFG FGKISGTACL TVYVQPIVSL HYKFSEDHLN ITCSATARPA PMVFWKVPRS GIENSTVTLS HPNGTTSVTS ILHIKDPKNQ VGKEVICQVL HLGTVTDFKQ TVNKG | |
Greater than 90% as determined by SDS-PAGE. |