missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Caspase-3 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
£320.73
Specifications
Gene ID (Entrez) | 836 |
---|---|
Molecular Weight (g/mol) | 17 kDa (subunit 1), 11 kDa (subunit 2) |
Name | Caspase-3 |
Quantity | 10μg |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18126490
|
R&D Systems™
707-C3-010/CF |
Supplied as a 0.2μm filtered solution in HEPES, NaCl, DTT and Sucrose. |
£320.73
10µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Human Caspase-3 Protein is derived from E. coli. The Recombinant Human Caspase-3 Protein has been validated for the following applications: Enzyme Activity.Specifications
836 | |
Caspase-3 | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. | |
Recombinant |
17 kDa (subunit 1), 11 kDa (subunit 2) | |
10μg | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |