missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Caspase-3 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Brand:  R&D Systems™ 707-C3-010/CF

Additional Details : Weight : 0.00970kg

 View more versions of this product

Product Code. 18126490

  • £320.73 / 10µg
Estimated Shipment: 04-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Human Caspase-3 Protein is derived from E. coli. The Recombinant Human Caspase-3 Protein has been validated for the following applications: Enzyme Activity.
Specifications

Specifications

Supplied as a 0.2μm filtered solution in HEPES, NaCl, DTT and Sucrose.
17 kDa (subunit 1), 11 kDa (subunit 2)
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening.
Recombinant
836
Caspase-3
E. coli-derived human Caspase-3 protein Ser29-Asp175 (subunit 1) & Ala183-His277 (Asp190Glu) (subunit 2)
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers