missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Calpastatin Protein
A DNA sequence encoding the Homo sapiens (Human) Calpastatin, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_001035905.1 |
---|---|
Gene ID (Entrez) | 831 |
Name | Calpastatin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15917599
|
enQuireBio™
QP5765-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15937599
|
enQuireBio™
QP5765-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15927599
|
enQuireBio™
QP5765-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15947599
|
enQuireBio™
QP5765-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15957599
|
enQuireBio™
QP5765-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15967599
|
enQuireBio™
QP5765-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001035905.1 | |
Calpastatin Protein | |
Not determined. | |
Full Length of isoform 4 | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
831 | |
Research Use Only | |
CAST | |
Recombinant Protein | |
E. coli | |
MNPTETKAVK TEPEKKSQST KPKSLPKQAS DTGSNDAHNK KAVSRSAEQQ PSEKSTEPKT KPQDMISAGG ESVAGITAIS GKPGDKKKEK KSLTPAVPVE SKPDKPSGKS GMDAALDDLI DTLGGPEETE EENTTYTGPE VSDPMSSTYI EELGKREVTI PPKYRELLAK KEGITGPPAD SSKPIGPDDA IDALSSDFTC GSPTAAGKKT EKEESTEVLK AQSAGTVRSA APPQEKKRKV EKDTMSDQAL EALSASLGTR QAEPELDLRS IKEVDEAKAK EEKLEKCGED DETIPSEYRL KPATDKDGKP LLPEPEEKPK PRSESELIDE LSEDFDRSEC KEKPSKPTEK TEESKAAAPA PVSEAVCRTS MCSIQSAPPE PATLKGTVPD DAVEALADSL GKKEADPEDG KPVMDKVKEK AKEEDREKLG EKEETIPPDY RLEEVKDKDG KPLLPKESKE QLPPMSEDFL LDALSEDFSG PQNASSLKFE DAKLAAAISE VVSQTPASTT QAGAPPRDTS QSDKDLDDAL DKLSDSLGQR QPDPDENKPM EDKVKEKAKA EHRDKLGERD DTIPPEYRHL LDDNGQDKPV KPPTKKSEDS KKPADDQDPI DALSGDLDSC PSTTETSQNT AKDKCKKAAS SSKAPKNGGK AKDSAKTTEE TSKPKDD | |
Greater than 90% as determined by SDS-PAGE. |