missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Calpastatin Protein
A DNA sequence encoding the Homo sapiens (Human) Calpastatin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5765-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001035905.1 | |
Calpastatin Protein | |
Research Use Only | |
CAST | |
Recombinant Protein | |
E. coli | |
MNPTETKAVK TEPEKKSQST KPKSLPKQAS DTGSNDAHNK KAVSRSAEQQ PSEKSTEPKT KPQDMISAGG ESVAGITAIS GKPGDKKKEK KSLTPAVPVE SKPDKPSGKS GMDAALDDLI DTLGGPEETE EENTTYTGPE VSDPMSSTYI EELGKREVTI PPKYRELLAK KEGITGPPAD SSKPIGPDDA IDALSSDFTC GSPTAAGKKT EKEESTEVLK AQSAGTVRSA APPQEKKRKV EKDTMSDQAL EALSASLGTR QAEPELDLRS IKEVDEAKAK EEKLEKCGED DETIPSEYRL KPATDKDGKP LLPEPEEKPK PRSESELIDE LSEDFDRSEC KEKPSKPTEK TEESKAAAPA PVSEAVCRTS MCSIQSAPPE PATLKGTVPD DAVEALADSL GKKEADPEDG KPVMDKVKEK AKEEDREKLG EKEETIPPDY RLEEVKDKDG KPLLPKESKE QLPPMSEDFL LDALSEDFSG PQNASSLKFE DAKLAAAISE VVSQTPASTT QAGAPPRDTS QSDKDLDDAL DKLSDSLGQR QPDPDENKPM EDKVKEKAKA EHRDKLGERD DTIPPEYRHL LDDNGQDKPV KPPTKKSEDS KKPADDQDPI DALSGDLDSC PSTTETSQNT AKDKCKKAAS SSKAPKNGGK AKDSAKTTEE TSKPKDD | |
Greater than 90% as determined by SDS-PAGE. |
831 | |
100 μg | |
Not determined. | |
Full Length of isoform 4 | |
Human | |
His | |
Tris-based buffer, 50% glycerol |