missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Calpain-2 catalytic subunit Protein
A DNA sequence encoding the Homo sapiens (Human) Calpain-2 catalytic subunit, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_001139540.1 |
---|---|
Gene ID (Entrez) | 824 |
Name | Calpain-2 catalytic subunit Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15957529
|
enQuireBio™
QP5756-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15977529
|
enQuireBio™
QP5756-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15967529
|
enQuireBio™
QP5756-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15987529
|
enQuireBio™
QP5756-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15997529
|
enQuireBio™
QP5756-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15907539
|
enQuireBio™
QP5756-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001139540.1 | |
Calpain-2 catalytic subunit Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
824 | |
Research Use Only | |
CAPN2 | |
Recombinant Protein | |
E. coli | |
SHDRAIKYLN QDYEALRNEC LEAGTLFQDP SFPAIPSALG FKELGPYSSK TRGIEWKRPT EICADPQFII GGATRTDICQ GALGDCWLLA AIASLTLNEE ILARVVPLNQ SFQENYAGIF HFQFWQYGEW VEVVVDDRLP TKDGELLFVH SAEGSEFWSA LLEKAYAKIN GCYEALSGGA TTEGFEDFTG GIAEWYELKK PPPNLFKIIQ KALQKGSLLG CSIDITSAAD SEAITFQKLV KGHAYSVTGA EEVESNGSLQ KLIRIRNPWG EVEWTGRWND NCPSWNTIDP EERERLTRRH EDGEFWMSFS DFLRHYSRLE ICNLTPDTLT SDTYKKWKLT KMDGNWRRGS TAGGCRNYPN TFWMNPQYLI KLEEEDEDEE DGESGCTFLV GLIQKHRRRQ RKMGEDMHTI GFGIYEVPEE LSGQTNIHLS KNFFLTNRAR ERSDTFINLR EVLNRFKLPP GEYILVPSTF EPNKDGDFCI RVFSEKKADY QAVDDEIEAN LEEFDISEDD IDDGFRRLFA QLAGEDAEIS AFELQTILRR VLAKRQDIKS DGFSIETCKI MVDMLDSDGS GKLGLKEFYI LWTKIQKYQK IYREIDVDRS GTMNSYEMRK ALEEAGFKMP CQLHQVIVAR FADDQLIIDF DNFVRCLVRL ETLFKIFKQL DPENTGTIEL DLISWLCFSV L | |
Greater than 80% as determined by SDS-PAGE. |