missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Calpain-2 catalytic subunit Protein
A DNA sequence encoding the Homo sapiens (Human) Calpain-2 catalytic subunit, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5756-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001139540.1 | |
Calpain-2 catalytic subunit Protein | |
Research Use Only | |
CAPN2 | |
Recombinant Protein | |
E. coli | |
SHDRAIKYLN QDYEALRNEC LEAGTLFQDP SFPAIPSALG FKELGPYSSK TRGIEWKRPT EICADPQFII GGATRTDICQ GALGDCWLLA AIASLTLNEE ILARVVPLNQ SFQENYAGIF HFQFWQYGEW VEVVVDDRLP TKDGELLFVH SAEGSEFWSA LLEKAYAKIN GCYEALSGGA TTEGFEDFTG GIAEWYELKK PPPNLFKIIQ KALQKGSLLG CSIDITSAAD SEAITFQKLV KGHAYSVTGA EEVESNGSLQ KLIRIRNPWG EVEWTGRWND NCPSWNTIDP EERERLTRRH EDGEFWMSFS DFLRHYSRLE ICNLTPDTLT SDTYKKWKLT KMDGNWRRGS TAGGCRNYPN TFWMNPQYLI KLEEEDEDEE DGESGCTFLV GLIQKHRRRQ RKMGEDMHTI GFGIYEVPEE LSGQTNIHLS KNFFLTNRAR ERSDTFINLR EVLNRFKLPP GEYILVPSTF EPNKDGDFCI RVFSEKKADY QAVDDEIEAN LEEFDISEDD IDDGFRRLFA QLAGEDAEIS AFELQTILRR VLAKRQDIKS DGFSIETCKI MVDMLDSDGS GKLGLKEFYI LWTKIQKYQK IYREIDVDRS GTMNSYEMRK ALEEAGFKMP CQLHQVIVAR FADDQLIIDF DNFVRCLVRL ETLFKIFKQL DPENTGTIEL DLISWLCFSV L | |
Greater than 80% as determined by SDS-PAGE. |
824 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |