missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BRAF Protein

A cDNA sequence encoding the BRAF was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11200-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15956458

  • £155.00 / 20µg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

BRAF Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSEFSEDRN RMKTLGRRDS SDDWEIPDGQ ITVGQRIGSG SFGTVYKGKW HGDVAVKMLN VTAPTPQQLQ AFKNEVGVLR KTRHVNILLF MGYSTKPQLA IVTQWCEGSS LYHHLHIIET KFEMIKLIDI ARQTAQGMDY LHAKSIIHRD LKSNNIFLHE DLTVKIGDFG LATVKSRWSG SHQFEQLSGS ILWMAPEVIR MQDKNPYSFQ SDVYAFGIVL YELMTGQLPY SNINNRDQII FMVGRGYLSP DLSKVRSNCP KAMKRLMAEC LKKKRDERPL FPQILASIEL LARSLPKIHR SASEPSLNRA GFQTEDFSLY ACASPKTPIQ AGGYGAFPVH
Greater than 80.0% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
BRAF protein solution (0.25 mg/ml) containing 20mM Tris-HCl (pH 8.0) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers