missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BRAF Protein
A cDNA sequence encoding the BRAF was constructed and used to recombinantly synthesize the protein.
£99.15 - £3965.00
Specifications
Name | BRAF Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15966458
|
enQuireBio™
QP11200-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15956458
|
enQuireBio™
QP11200-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15946458
|
enQuireBio™
QP11200-1MG |
1 mg |
£3965.00
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
BRAF Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
BRAF protein solution (0.25 mg/ml) containing 20mM Tris-HCl (pH 8.0) and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSEFSEDRN RMKTLGRRDS SDDWEIPDGQ ITVGQRIGSG SFGTVYKGKW HGDVAVKMLN VTAPTPQQLQ AFKNEVGVLR KTRHVNILLF MGYSTKPQLA IVTQWCEGSS LYHHLHIIET KFEMIKLIDI ARQTAQGMDY LHAKSIIHRD LKSNNIFLHE DLTVKIGDFG LATVKSRWSG SHQFEQLSGS ILWMAPEVIR MQDKNPYSFQ SDVYAFGIVL YELMTGQLPY SNINNRDQII FMVGRGYLSP DLSKVRSNCP KAMKRLMAEC LKKKRDERPL FPQILASIEL LARSLPKIHR SASEPSLNRA GFQTEDFSLY ACASPKTPIQ AGGYGAFPVH | |
Greater than 80.0% as determined by SDS-PAGE. |