missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human BMP-2/BMP-7 Heterodimer Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£490.27
Specifications
Gene ID (Entrez) | 650 |
---|---|
Molecular Weight (g/mol) | 12.9 kDa (BMP-2), 15.8 kDa (BMP-7) |
Name | BMP-2/BMP-7 Heterodimer |
Quantity | 10μg |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70 °C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening. |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18318947
|
R&D Systems™
3229-BM-010 |
£490.27
10µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18178160
|
R&D Systems™
3229-BM-010/CF |
Supplied as a 0.2 μm filtered solution in Acetonitrile and TFA. |
£490.27
10µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Human BMP-2/BMP-7 Heterodimer Protein is derived from E. coli. The Recombinant Human BMP-2/BMP-7 Heterodimer Protein has been validated for the following applications: Bioactivity.Specifications
650 | |
BMP-2/BMP-7 Heterodimer | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70 °C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening. | |
Unconjugated | |
>97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
12.9 kDa (BMP-2), 15.8 kDa (BMP-7) | |
10μg | |
Disulfide-linked heterodimer | |
Recombinant |