missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human BMP-2/BMP-7 Heterodimer Protein
Click to view available options
Formulation:
Supplied as 0.2 μm filtered solution in 30%(v/v) Acetonitrile and 0.1%(v/v) TFA with 50 μg BSA / 1 μg as a carrier protein.
Supplied as a 0.2 μm filtered solution in Acetonitrile and TFA.
Unit Size:
10µg
Description
Measured by its ability to induce alkaline phosphatase production by ATDC5 mouse chondrogenic cells. Nakamura, K. et al. (1999) Exp. Cell Res. 250:351. The ED50 for this effect is 15.0-120 ng/mL.
Specifications
Specifications
| Formulation | Supplied as a 0.2 μm filtered solution in Acetonitrile and TFA. |
| Gene ID (Entrez) | 650 |
| Molecular Weight (g/mol) | 12.9 kDa (BMP-2), 15.8 kDa (BMP-7) |
| Name | BMP-2/BMP-7 Heterodimer |
| Quantity | 10 μg |
| Source | E. coli-derived human BMP-2/BMP-7 Heterodimer protein, (N-terminus) Human BMP-2 (Ala284 – Arg396) Accession &Num; NP_001191 Human BMP-7 (Ser293 – His431) Accession &Num; NP_001710 (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70 °C as supplied. 1 month, 2°C to 8°C under sterile conditions after opening. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Structural Form | Disulfide-linked heterodimer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction