missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human BDNF Protein

Product Code. 18797206
Change view
Click to view available options
Quantity:
1 mg
10 μg
250 μg
50 μg
Unit Size:
10µg
1mg
250µg
50µg
Product Code. Quantity unitSize
18797206 10 μg 10µg
18717118 50 μg 50µg
18745596 250 μg 250µg
18748866 1 mg 1mg
4 options
This item is not returnable. View return policy

Product Code. 18797206

Brand: R&D Systems™ 248BDB010/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human BDNF Protein is derived from Sf 21 (baculovirus). The Recombinant Human BDNF Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P23560
For Use With (Application) Bioactivity
Gene ID (Entrez) 627
Molecular Weight (g/mol) observed_molecular_weight: 13-14 kDa, under reducing conditions.
Quantity 10 μg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived BDNF protein His129-Arg247 Accession &Num; P23560 100% sequence homology with Mouse,Rat,Canine,Equine and all other mammalian proteins examined.
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Specific Reactivity BDNF
Conjugate Unconjugated
Species Human, Mouse, Rat, Canine, Equine
Recombinant Recombinant
Reconstitution Reconstitute at 250 μg/mL in water.
Buffer Lyophilized from a 0.2 μm filtered solution in Sodium Citrate and NaCl.
Purity or Quality Grade >97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie« Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.